Anti-RNF5 Mouse Monoclonal Antibody (DyLight® 680) [clone: LHM 2]
NOVUNB100-2688FR
- Antibody type:Primary
- Antigen name:chondroitin sulfate proteoglycan 4
- Antigen symbol:CSPG4
- Clonality:Monoclonal
- Clone:LHM 2
- Conjugation:DyLight® 680
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Gene ID:1464
- Antigen synonyms:MEL-CSPG|HMW-MAA|MCSP|MCSPG|NG2|MSK16
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 Tests
- Pk:100 Tests
Type: Primary
Antigen: RNF5 (ring finger protein 5, E3 ubiquitin protein ligase)
Clonality: monoclonal
Clone: LHM 2
Conjugation: DyLight® 680
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human