Anti-DNMT3A Rabbit Polyclonal Antibody
Type: Primary
Antigen: DNMT3A (DNA (cytosine-5-)-methyltransferase 3 alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
- Catalog No:
- NOVUNB100-265
- NOVUNB100-265SS
- Antigen symbol:
- DNMT3A
- DNMT3A
- Antigen name:
- DNA (cytosine-5-)-methyltransferase 3 alpha
- DNA (cytosine-5-)-methyltransferase 3 alpha
- Antigen synonymer:
- 2.1.1.37|DNA (cytosine-5)-methyltransferase 3A|DNA MTase HsaIIIA|M.HsaIIIA|Dnmt3a|DNA methyltransferase HsaIIIA
- 2.1.1.37|DNA (cytosine-5)-methyltransferase 3A|DNA MTase HsaIIIA|M.HsaIIIA|Dnmt3a|DNA methyltransferase HsaIIIA
- Antistof type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Konjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Vært:
- Rabbit
- Rabbit
- Gen ID:
- 1788
- 1788
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunFluorescens:
- Yes
- Yes
- Opbevaringstemperatur:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together