Order Entry
Denmark
ContactUsLinkComponent
Anti-VEGFA Rabbit Polyclonal Antibody
Anti-VEGFA Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals
Certificates
undefined
Anti-VEGFA Rabbit Polyclonal Antibody
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNB100-2381
  • NOVUNB100-2381SS
  • Antigen symbol:
  • VEGFA
  • VEGFA
  • Antigen name:
  • Vascular endothelial growth factor A
  • Vascular endothelial growth factor A
  • Antigen synonyms:
  • VEGF|VEGF-A,|Vascular permeability factor|Vascular Endothelial Growth Factor|Vascular endothelial growth factor A
  • VEGF|VEGF-A,|Vascular permeability factor|Vascular Endothelial Growth Factor|Vascular endothelial growth factor A
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 7422
  • 7422
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together