Order Entry
Denmark
ContactUsLinkComponent
Anti-HMGB1 Rabbit Polyclonal Antibody (DyLight® 550)
Anti-HMGB1 Rabbit Polyclonal Antibody (DyLight® 550)
Catalog # NOVUNB100-2322R
Supplier:  Novus Biologicals
undefined
Anti-HMGB1 Rabbit Polyclonal Antibody (DyLight® 550)
Catalog # NOVUNB100-2322R
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    High-mobility Group Box 1
  • Antigen symbol:
    HMGB1
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 550
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    3146
  • Antigen synonyms:
    high-mobility group box 1|RP11-550P23.1|high mobility group protein 1|Amphoterin|SBP-1|high-mobility group (nonhistone chromosomal) protein 1|high mobility group protein B1|HMG3|Sulfoglucuronyl carbohydrate binding protein|HMG1|HMG-1
  • Storage buffer:
    50 mM Sodium Borate
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    0,1 mL

Specifications

About this item

Rabbit Polyclonal HMGB1 Antibody [DyLight 550]. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Rat, Bovine, Canine.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:100-1:250

Type: Primary
Antigen: HMGB1
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 550
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human