Anti-MAP1LC3A Rabbit Polyclonal Antibody
Type: Primary
Antigen: MAP1LC3A (microtubule-associated protein 1 light chain 3 alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
- Catalog No:
- NOVUNB100-2220
- NOVUNB100-2220SS
- Antigen symbol:
- LC3
- LC3
- Antigen name:
- 1A/1B-light chain 3
- 1A/1B-light chain 3
- Antigen synonyms:
- MAP1A/1BLC3|MAP1LC3B-a|ATG8F|LC3B
- MAP1A/1BLC3|MAP1LC3B-a|ATG8F|LC3B
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 81631
- 81631
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together