Order Entry
Denmark
ContactUsLinkComponent
Anti-XRCC3 Rabbit Polyclonal Antibody (DyLight® 650)
Anti-XRCC3 Rabbit Polyclonal Antibody (DyLight® 650)
Catalog # NOVUNB100-165C
Supplier:  Novus Biologicals
undefined
Anti-XRCC3 Rabbit Polyclonal Antibody (DyLight® 650)
Catalog # NOVUNB100-165C
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    X-ray Repair Complementing Defective Repair in Chinese Hamster cells 3
  • Antigen symbol:
    XRCC3
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 650
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    7517
  • Antigen synonyms:
    X ray repair complementing defective repair in Chinese hamster cells 3|CMM6|X ray repair cross complementing protein 3|DNA repair protein XRCC3
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: XRCC3 (X-ray repair complementing defective repair in Chinese hamster cells 3)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 650
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human