Anti-CITED2 Mouse monoclonal antibody unconjugated [clone: JA22]
Type: Primary
Antigen: CITED2 (Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain, 2)
Clonality: monoclonal
Clone: JA22
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse
- Catalog No:
- NOVUNB100-136SS
- NOVUNB100-136
- Antigen symbol:
- CITED2
- CITED2
- Antigen name:
- Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal Domain, 2
- Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal Domain, 2
- Antigen synonyms:
- P35SRJ|MRG1|Cbp/p300-interacting transactivator with Glu/Asp-rich carboxy-terminal domain 2
- P35SRJ|MRG1|Cbp/p300-interacting transactivator with Glu/Asp-rich carboxy-terminal domain 2
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- JA22
- JA22
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 10370
- 10370
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together