Anti-HIF1A Mouse Monoclonal Antibody [clone: H1alpha 67-7]
Type: Primary
Antigen: HIF1A (hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor))
Clonality: monoclonal
Clone: H1alpha 67-7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
- Catalog No:
- NOVUNB100-130SS
- NOVUNB100-130
- Antigen symbol:
- HIF1A
- HIF1A
- Antigen name:
- HIF1 alpha
- HIF1 alpha
- Antigen synonyms:
- PASD8|HIF-1alpha|HIF1-ALPHA|HIF-1A|HIF1|MOP1|HIF-1-alpha|bHLHe78
- PASD8|HIF-1alpha|HIF1-ALPHA|HIF-1A|HIF1|MOP1|HIF-1-alpha|bHLHe78
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- H1alpha 67-7
- H1alpha 67-7
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 3091
- 3091
- Isotype:
- IgG2b
- IgG2b
- Western blot:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together