Order Entry
Denmark
ContactUsLinkComponent
Anti-NTHL1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Anti-NTHL1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # NOVUNB100-108F
Supplier:  Novus Biologicals
Anti-NTHL1 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Catalog # NOVUNB100-108F
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    nth-like DNA glycosylase 1
  • Antigen symbol:
    NTHL1
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Gene ID:
    4913
  • Antigen synonyms:
    NTH1|FAP3|OCTS3|hNTH1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    200 μL
  • Pk:
    0,2 mL

Specifications

About this item

Type: Primary
Antigen: NTHL1 (nth endonuclease III-like 1 (E. coli))
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat