Anti-APEX1 Rabbit Polyclonal Antibody (Biotin)
Catalog # NOVUNB100-101B
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:APEX nuclease (multifunctional DNA repair enzyme) 1
- Antigen symbol:APEX1
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Gene ID:328
- Antigen synonyms:Protein REF-1|APEX|DNA-(apurinic or apyrimidinic site) lyase|APEN|REF1|APE|APEX nuclease|APX|AP endonuclease 1|Apurinic-apyrimidinic endonuclease 1|APEX1|HAP1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: APEX1 (APEX nuclease (multifunctional DNA repair enzyme) 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: