Anti-Transglutaminase 2 Rabbit Polyclonal Antibody
Catalog # BSBTPA2153
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Transglutaminase 2
- Antigen symbol:TG2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:tTG|TGM2_HUMAN|Transglutaminase 2 C polypeptide|TGase2|Tissue type transglutaminase|H POLYPEPTIDE|Tissue transglutaminase|TG 2|G protein alpha subunit Gh class|TG2|C polypeptide|TGM2|Gh CLASS G ALPHA h|GNAH|G[a]h|TGC|Guanine nucleotide binding protein H polypeptide|ALPHA SUBUNIT|TGC GUANINE NUCLEOTIDE BINDING PROTEIN|Transglutaminase-2|TGase C|TGase H|TgaseII|Transglutaminase H|TGM 2|G alpha h|tTGas.|Transglutaminase 2|Protein glutamine gamma glutamyltransferase|Protein-glutamine gamma-glutamyltransferase 2|TGase-2|GNAH G PROTEIN|Protein glutamine gamma glutamyltransferase 2|Transglutaminase C
- Amino acid number:399 - 418
- Sequence:ADVVDWIQQDDGSVHKSINR
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Transglutaminase 2 (399-418aa ADVVDWIQQDDGSVHKSINR), different from the related mouse sequence by three amino acids, and from the related rat sequence by four amino acids
- Purification:Purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Protein-glutamine gamma-glutamyltransferase 2(TGM2) detection. Tested with WB, IHC-P, ICC in Human, Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Transglutaminase 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat