Anti-ABCG4 Rabbit Polyclonal Antibody
BSBTPA2067
- Antibody type:Primary
- Antigen name:ATP-binding cassette, Subfamily G (WHITE), Member 4
- Antigen symbol:ABCG4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:ATP-binding cassette sub-family G member 4|WHITE2
- Amino acid number:327 - 341
- Sequence:AVQNGLCAMAEKKSS
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human ABCG4 (327-341aa AVQNGLCAMAEKKSS), different from the related mouse and rat sequences by one amino acid
- Purification:Purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for ATP-binding cassette sub-family G member 4(ABCG4) detection. Tested with WB in Human, Mouse, Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: ABCG4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat