Order Entry
Denmark
ContactUsLinkComponent
 
Anti-STAT5b Rabbit Polyclonal Antibody
  BSBTPA1841
undefined
Anti-STAT5b Rabbit Polyclonal Antibody
  BSBTPA1841

 

  • Antibody type:
    Primary
  • Antigen name:
    Signal transducer and activator of transcription 5B
  • Antigen symbol:
    STAT5B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilised
  • Antigen synonyms:
    STAT5|Signal transducer and activator of transcription 5B|Transcription factor STAT5B|Stat5b.|STA5B_HUMAN
  • Amino acid number:
    767 - 787
  • Sequence:
    RRVEELLGRPMDSQWIPHAQS
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human STAT5b (767-787aa RRVEELLGRPMDSQWIPHAQS), identical to the related rat and mouse sequences.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Rabbit IgG polyclonal antibody for Signal transducer and activator of transcription 5B (STAT5B) detection. Tested with WB, IHC-P in Human; Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: STAT5b
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat