Order Entry
Denmark
ContactUsLinkComponent
Anti-Kallikrein 10 Rabbit Polyclonal Antibody
Anti-Kallikrein 10 Rabbit Polyclonal Antibody
Catalog # BSBTPA1821
Supplier:  Boster Bio
undefined
Anti-Kallikrein 10 Rabbit Polyclonal Antibody
Catalog # BSBTPA1821
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Kallikrein 10
  • Antigen symbol:
    KLK10
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilised
  • Amino acid number:
    252 - 265
  • Sequence:
    SAQHPAVYTQICKY
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human Kallikrein 10 (252-265aa SAQHPAVYTQICKY).
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Kallikrein-10 (KLK10) detection. Tested with WB in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Kallikrein 10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human