Anti-CBL Rabbit Polyclonal Antibody
Catalog # BSBTPA1800
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Cbl proto-oncogene, E3 Ubiquitin Protein Ligase
- Antigen symbol:CBL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:RING finger protein 57|SH3-binding protein CBL-C|RNF57|CBLC|CBL-3|Signal transduction protein CBL-C
- Amino acid number:863 - 877
- Sequence:NLMSQGYSYQDIQKA
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human CBL (863-877aa NLMSQGYSYQDIQKA), identical to the related rat and mouse sequences.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for E3 ubiquitin-protein ligase CBL (CBL) detection. Tested with WB in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: CBL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat