Anti-Kallikrein 5 Rabbit Polyclonal Antibody
BSBTPA1630
- Antibody type:Primary
- Antigen name:Kallikrein 5
- Antigen symbol:KLK5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:Stratum corneum tryptic enzyme|Kallikrein-like protein 2|SCTE|KLK-L2
- Amino acid number:278 - 293
- Sequence:NLCEFVKWIKDTMNSN
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of mouse Kallikrein 5 (278-293aa NLCEFVKWIKDTMNSN), different from the related rat sequence by three amino acids.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for Kallikrein related-peptidase 5/Protein Klk5(KLK5) detection. Tested with WB, IHC-P in Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Kallikrein 5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat