Order Entry
Denmark
ContactUsLinkComponent
 
Anti-FOXP3 Rabbit Polyclonal Antibody
  BSBTPA1577
undefined
Anti-FOXP3 Rabbit Polyclonal Antibody
  BSBTPA1577

 

  • Antibody type:
    Primary
  • Antigen name:
    Forkhead Box P3
  • Antigen symbol:
    FOXP3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    AIID|PIDX|JM2|DIETER|XPID|Scurfin|IPEX
  • Amino acid number:
    321 - 337
  • Sequence:
    FPEFLHNMDYFKFHNMR
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence in the middle region of human FOXP3 (321-337aa FPEFLHNMDYFKFHNMR), different from the related mouse sequence by two amino acids.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Rabbit IgG polyclonal antibody for Forkhead box protein P3(FOXP3) detection. Tested with WB in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: FOXP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human