Anti-c-Myc Rabbit Polyclonal Antibody
BSBTPA1555
- Antibody type:Primary
- Antigen symbol:c-Myc
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:421 - 439
- Sequence:RKRREQLKHKLEQLRNSCA
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human c-Myc (421-439aa RKRREQLKHKLEQLRNSCA), different from the related mouse and rat sequences by one amino acid.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for Myc proto-oncogene protein(MYC) detection. Tested with WB in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: c-Myc
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat