Anti-AIMP2/p38 Rabbit Polyclonal Antibody
BSBTPA1481
- Antibody type:Primary
- Antigen name:AIMP2/p38
- Antigen symbol:AIMP2/p38
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:p38
- Amino acid number:298 - 320
- Sequence:NVQRWMRSCENLAPFNTALKLLK
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human AIMP2/p38 (298-320aa NVQRWMRSCENLAPFNTALKLLK), different from the related rat and mouse sequences by three amino acids.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for Aminoacyl tRNA synthase complex-interacting multifunctional protein 2(AIMP2) detection. Tested with WB, IHC-P in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: AIMP2/p38
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat