Anti-Angiopoietin-like 4 Rabbit Polyclonal Antibody
Catalog # BSBTPA1435
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Angiopoietin-like 4
- Antigen symbol:ANGPTL4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:PSEC0166|ANGPTL4|ARP4|HFARP|pp1158|NL2|FIAF|PGAR|UNQ171/PRO197|ANGPTL2
- Amino acid number:349 - 367
- Sequence:WWFGTCSHSNLNGQYFRSI
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Angiopoietin-like 4 (349-367aa WWFGTCSHSNLNGQYFRSI), different from the related rat and mouse sequences by one amino acid.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Angiopoietin-related protein 4(ANGPTL4) detection. Tested with WB in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Angiopoietin-like 4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat