Order Entry
Denmark
ContactUsLinkComponent
 
Anti-Cdk4 Rabbit Polyclonal Antibody
Catalog # BSBTPA1428-1
Supplier:  Boster Bio
Anti-Cdk4 Rabbit Polyclonal Antibody
Catalog # BSBTPA1428-1
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cyclin-dependent Kinase 4
  • Antigen symbol:
    CDK4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    CMM3|MGC14458|PSK-J3|Melanoma Cutaneous Malignant 3|Cell Division Kinase 4|Cell Division Protein Kinase 4
  • Amino acid number:
    283 - 303
  • Sequence:
    RISAFRALQHSYLHKEESDAE
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of mouse Cdk4 (283-303aa RISAFRALQHSYLHKEESDAE), different from the related rat sequence by one amino acid, and from the related human sequence by four amino acids.
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Cyclin-dependent kinase 4(CDK4) detection. Tested with WB in Human; Mouse; Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Cdk4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat