Order Entry
Denmark
ContactUsLinkComponent
Anti-c-Fos Rabbit Polyclonal Antibody
Anti-c-Fos Rabbit Polyclonal Antibody
Catalog # BSBTPA1318
Supplier:  Boster Bio
undefined
Anti-c-Fos Rabbit Polyclonal Antibody
Catalog # BSBTPA1318
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    c-Fos
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Lyophilised
  • Amino acid number:
    170 - 187
  • Sequence:
    DQLEDEKSALQTEIANLL
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence in the middle region of human c-Fos (170-187aa DQLEDEKSALQTEIANLL), identical to the related rat and mouse sequences
  • Purification:
    purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Proto-oncogene c-Fos (FOS) detection. Tested with WB in Human;Mouse;Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: c-Fos
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat