Anti-MMP16 Rabbit Polyclonal Antibody
Catalog # BSBTPA1123
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Matrix Metalloproteinase 16
- Antigen symbol:MMP16
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:DKFZp761D112|MT-MMP2|MMP-X2|MT-MMP3|MT3-MMP|C8orf57
- Amino acid number:582 - 598
- Sequence:YTVFQFKRKGTPRHILY
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human MMP16 (582-598aa, YTVFQFKRKGTPRHILY), identical to the related rat and mouse sequences
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Matrix metalloproteinase-16 (MMP16) detection. Tested with WB, IHC-P, IHC-F in Human;Mouse;Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: MMP16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat