Anti-alpha-Smooth Muscle Actin Mouse Monoclonal Antibody [clone: 1A4]
Catalog # BSBTMA1106
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Smooth Muscle Actin
- Antigen symbol:ACTA2
- Clonality:Monoclonal
- Clone:1A4
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Ascites
- Form:Lyophilised
- Antigen synonyms:AAT6|alpha 2 actin|MYMY5|aortic smooth muscle|cell growth-inhibiting gene 46 protein|alpha-cardiac actin|alpha-actin-2|actin|GIG46|growth-inhibiting gene 46|ACTSA
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:N-terminal synthetic decapeptide of alpha-smooth muscle actin
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Mouse IgG monoclonal antibody for alpha-Smooth Muscle Actin, actin, alpha 2, smooth muscle, aorta (ACTA2) detection. Tested with WB, IHC-P, IHC-F in Human;mouse;rat. No cross reactivity with other proteins.
Reconstitute with 1,2% sodium acetate or neutral PBS.
If 1 ml of PBS is used, the antibody concentration will be 100 µg/ml.
Type: Primary
Antigen: alpha-Smooth Muscle Actin
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat