Order Entry
Denmark
ContactUsLinkComponent
Anti-Cathepsin L Mouse Monoclonal Antibody [clone: CP-L14]
Anti-Cathepsin L Mouse Monoclonal Antibody [clone: CP-L14]
Catalog # BSBTMA1014
Supplier:  Boster Bio
Anti-Cathepsin L Mouse Monoclonal Antibody [clone: CP-L14]
Catalog # BSBTMA1014
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cathepsin L
  • Antigen symbol:
    CTSL
  • Clonality:
    Monoclonal
  • Clone:
    CP-L14
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Ascites
  • Form:
    Lyophilized
  • Antigen synonyms:
    nkt|MEP|CTSL1|CATL|fs
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    Procathepsin L isolated from the human lung cancer cell line EPLC 32M1.
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Type: Primary
Antigen: Cathepsin L
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat