Anti-C Reactive Protein Mouse Monoclonal Antibody [clone: C2]
Catalog # ABCAAB136176-1
Supplier: Abcam
Specifications
- Antibody type:Primary
- Antigen name:C Reactive Protein
- Antigen symbol:CRP
- Clonality:Monoclonal
- Clone:C2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 mg
- Pack type:Vial
- Pk:1 mg
Specifications
About this item
Mouse monoclonal [C2] to C Reactive Protein.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: C Reactive Protein
Clonality: Monoclonal
Clone: C2
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human