Order Entry
Denmark
ContactUsLinkComponent
Anti-CXCL7/PBP Rabbit Monoclonal Antibody [clone: EPR7767]
Anti-CXCL7/PBP Rabbit Monoclonal Antibody [clone: EPR7767]
Supplier:  Abcam
New Product
Certificates
undefined
Anti-CXCL7/PBP Rabbit Monoclonal Antibody [clone: EPR7767]
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCAAB133252-100
  • ABCAAB133252-1ML
  • Antigen symbol:
  • NAP1L4
  • Antigen name:
  • nucleosome assembly Protein 1-like 4
  • Antibody type:
  • Primary
  • Clonality:
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Clone:
  • EPR7767
  • Host:
  • Rabbit
  • Isotype:
  • IgG
  • Western blot:
  • Yes
  • Storage temperature:
  • Cool
  • Shipping temperature:
  • Ice

Specifications

Frequently Bought Together