Anti-ZNF704 Rabbit Polyclonal Antibody
Catalog # ABCAAB121804-100
Supplier: Abcam
Specifications
- Antibody type:Primary
- Antigen name:zinc finger protein 704
- Antigen symbol:ZNF704
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Seq:1286
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal to ZNF704.
Caution:
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: ZNF704
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human