Order Entry
Denmark
ContactUsLinkComponent
Anti-5-hydroxymethylcytosine (5-hmC) Rat Monoclonal Antibody [clone: AB3/63.3]
Anti-5-hydroxymethylcytosine (5-hmC) Rat Monoclonal Antibody [clone: AB3/63.3]
Catalog # ABCAAB106918-100
Supplier:  Abcam
undefined
Anti-5-hydroxymethylcytosine (5-hmC) Rat Monoclonal Antibody [clone: AB3/63.3]
Catalog # ABCAAB106918-100
Supplier:  Abcam

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    5-Hydroxymethyl Cytidine
  • Clonality:
    Monoclonal
  • Clone:
    AB3/63.3
  • Conjugation:
    Unconjugated
  • Host:
    Rat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pack type:
    Vial
  • Pk:
    100 µG

Specifications

About this item

Rat monoclonal [AB3/63.3] to 5-hydroxymethylcytosine (5-hmC) - ChIP Grade.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: 5-hydroxymethylcytosine (5-hmC)
Clonality: Monoclonal
Clone: AB3/63.3
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: