Order Entry
Denmark
ContactUsLinkComponent
 
Anti-NMDAR2B Rabbit Monoclonal Antibody [clone: EP1858Y]
 :  Abcam
New Product
 
undefined
Anti-NMDAR2B Rabbit Monoclonal Antibody [clone: EP1858Y]
 :  Abcam
New Product

 

  • Catalog No:
  • ABCAAB81271-1ML
  • ABCAAB81271-100
  • ABCAAB81271-20UL
  • Antigen symbol:
  • GRIN2B
  • Antigen name:
  • Glutamate receptor, ionotropic, N-methyl D-aspartate 2B
  • Antibody type:
  • Primary
  • Clonality:
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Clone:
  • EPR1858Y
  • Host:
  • Rabbit
  • Isotype:
  • IgG
  • Western blot:
  • Yes
  • ImmunoChemistry:
  • Yes
  • Flow cytometry:
  • Yes
  • Storage temperature:
  • Cool
  • Shipping temperature:
  • Ice

 

Frequently Bought Together