Order Entry
Denmark
ContactUsLinkComponent
Anti-Smad1+Smad2+Smad3+SMAD5 (phospho S423 + S425) Rabbit Monoclonal Antibody [clone: EP823Y]
Anti-Smad1+Smad2+Smad3+SMAD5 (phospho S423 + S425) Rabbit Monoclonal Antibody [clone: EP823Y]
Supplier:  Abcam
New Product
Certificates
undefined
Anti-Smad1+Smad2+Smad3+SMAD5 (phospho S423 + S425) Rabbit Monoclonal Antibody [clone: EP823Y]
Supplier:  Abcam
New Product

Specifications

  • Catalog No:
  • ABCAAB52903-100
  • ABCAAB52903-20UL
  • ABCAAB52903-1ML
  • Antigen symbol:
  • SMAD family member 3
  • Antibody type:
  • Primary
  • Clonality:
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Clone:
  • EP823Y
  • Host:
  • Rabbit
  • Isotype:
  • IgG
  • Western blot:
  • Yes
  • ELISA:
  • Yes
  • ImmunoChemistry:
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Storage temperature:
  • Cool
  • Shipping temperature:
  • Ice

Specifications

Frequently Bought Together