- Antibody type:Primary
- Antigen name:Cyclin-dependent kinase inhibitor 2B (p15, inhibits CDK4)
- Antigen symbol:CDKN2B
- Clonality:Monoclonal
- Clone:DCS-114
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:250 µl
- Pk:250 µl
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CDKN2B (cyclin-dependent kinase inhibitor 2B (p15, inhibits CDK4))
Clonality: monoclonal
Clone: DCS-114
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human