Anti-JMY Mouse Monoclonal Antibody [clone: 2D7]
Catalog # ABNOH00133746-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Junction-Mediating And Regulatory Protein
- Antigen symbol:JMY
- Clonality:Monoclonal
- Clone:2D7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:133746
- Antigen synonyms:WHDC1L3|P53 Cofactor|Junction-mediating and -regulatory protein|WAS Protein Homology Region 2 Domain Containing 1-Like 3|Junction Mediating And Regulatory Protein|JMY|WHAMM2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: JMY
Clonality: Monoclonal
Clone: 2D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human