Anti-PRRT2 Mouse Polyclonal Antibody
Catalog # ABNOH00112476-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Proline-rich Transmembrane Protein 2
- Antigen symbol:PRRT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:112476
- Antigen synonyms:Dispanin subfamily B member 3|ICCA|Proline-rich transmembrane protein 2|FICCA|PKC|EKD1|DYT10|PRRT2|DSPB3|IFITMD1|BFIS2|BFIC2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: PRRT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human