Anti-GAL3ST4 Mouse Polyclonal Antibody
ABNOH00079690-B01P
: Abnova
- Antibody type:Primary
- Antigen name:Galactose-3-O-sulfotransferase 4
- Antigen symbol:GAL3ST4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:79690
- Antigen synonyms:GAL3ST-4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: GAL3ST4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human