Anti-SIX4 Mouse Monoclonal Antibody [clone: 7F1]
ABNOH00051804-M02
: Abnova
- Antibody type:Primary
- Antigen name:SIX Homeobox 4
- Clonality:Monoclonal
- Clone:7F1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:51804
- Antigen synonyms:AREC3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: SIX4
Clonality: Monoclonal
Clone: 7F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human