Anti-KCNIP3 Rabbit Polyclonal Antibody
Catalog # ABNOH00030818-D01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Kv Channel Interacting Protein 3
- Antigen symbol:KCNIP3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:30818
- Antigen synonyms:DREAM|CSEN|KCHIP3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: KCNIP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse