Order Entry
Denmark
ContactUsLinkComponent
Anti-MGRN1 Mouse Monoclonal Antibody [clone: 3E1]
Anti-MGRN1 Mouse Monoclonal Antibody [clone: 3E1]
Catalog # ABNOH00023295-M07
Supplier:  Abnova
undefined
Anti-MGRN1 Mouse Monoclonal Antibody [clone: 3E1]
Catalog # ABNOH00023295-M07
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Mahogunin RING Finger Protein 1
  • Antigen symbol:
    MGRN1
  • Clonality:
    Monoclonal
  • Clone:
    3E1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    23295
  • Antigen synonyms:
    KIAA0544|RING Finger Protein 156|Mahogunin|Ring Finger 1|RNF156
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: MGRN1
Clonality: Monoclonal
Clone: 3E1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human