Anti-GAL3ST1 Rabbit Polyclonal Antibody
100 µg
Catalog # ABNOH00009514-D01P
Supplier: Abnova
Jump to:
Product Details & Documents
Type: Primary
Antigen: GAL3ST1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Documents
TDS_ABNOH00009514-D01P.pdf
Datasheet
Specifications
- Size:100 µg
- Western blot:Yes
- Clonality:Polyclonal
- Antibody type:Primary
- Storage temperature:Cool
- Host:Rabbit
- Cross adsorption:No
- Shipping temperature:Ice
- Antigen name:Galactose-3-O-sulfotransferase 1
- Cat. No.:ABNOH00009514-D01P
- Reactivity:Human
- Gene ID:9514
- Antigen synonyms:CST
- Conjugation:Unconjugated
- Antigen symbol:GAL3ST1
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



