Anti-CNBP Mouse Polyclonal Antibody
Catalog # ABNOH00007555-A01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:CCHC-type Zinc Finger, nucleic acid binding protein
- Antigen symbol:CNBP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:7555
- Antigen synonyms:CNBP1|RNF163|DM2|PROMM|ZCCHC22|ZNF9
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: CNBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human