Anti-ZIC3 Mouse Monoclonal Antibody [clone: 4F7]
Catalog # ABNOH00007547-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Zinc Finger Protein of the cerebellum 3
- Antigen symbol:ZIC3
- Clonality:Monoclonal
- Clone:4F7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:7547
- Antigen synonyms:HTX|VACTERLX|ZNF203|HTX1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: ZIC3
Clonality: Monoclonal
Clone: 4F7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human