Anti-WRN Mouse Monoclonal Antibody [clone: 3C11]
Catalog # ABNOH00007486-M09
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Werner syndrome, RecQ helicase-like
- Antigen symbol:WRN
- Clonality:Monoclonal
- Clone:3C11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:7486
- Antigen synonyms:RECQL3|RECQ3|RECQL2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: WRN
Clonality: Monoclonal
Clone: 3C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human