Order Entry
Denmark
ContactUsLinkComponent
Anti-TEC Mouse Monoclonal Antibody [clone: 1C11]
Anti-TEC Mouse Monoclonal Antibody [clone: 1C11]
Catalog # ABNOH00007006-M01
Supplier:  Abnova
Anti-TEC Mouse Monoclonal Antibody [clone: 1C11]
Catalog # ABNOH00007006-M01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Tec protein tyrosine kinase
  • Antigen symbol:
    TEC
  • Clonality:
    Monoclonal
  • Clone:
    1C11
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Gene ID:
    7006
  • Antigen synonyms:
    PSCTK4|MGC126762|MGC126760|TEC
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: TEC
Clonality: Monoclonal
Clone: 1C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human