Anti-SMO Mouse Monoclonal Antibody [clone: 3B1]
ABNOH00006608-M08
: Abnova
- Antibody type:Primary
- Antigen name:Smoothened protein
- Antigen symbol:SMO
- Clonality:Monoclonal
- Clone:3B1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:6608
- Antigen synonyms:Gx|SMOH|FZD11|smoothened|frizzled class receptor
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: SMO
Clonality: Monoclonal
Clone: 3B1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human