Anti-RGL2 Mouse Monoclonal Antibody [clone: 4D10]
ABNOH00005863-M02
: Abnova
- Antibody type:Primary
- Antigen name:Ral Guanine Nucleotide Dissociation Stimulator-like 2
- Antigen symbol:RGL2
- Clonality:Monoclonal
- Clone:4D10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5863
- Antigen synonyms:HKE1.5|RAB2L|KE1.5
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: RGL2
Clonality: Monoclonal
Clone: 4D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human