Anti-HLF Mouse Monoclonal Antibody [clone: 1F12-1C4]
ABNOH00003131-M01
: Abnova
- Antibody type:Primary
- Antigen name:Hepatic Leukemia Factor
- Antigen symbol:HLF
- Clonality:Monoclonal
- Clone:1F12-1C4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:3131
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: HLF
Clonality: Monoclonal
Clone: 1F12-1C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human