Anti-GALT Mouse Monoclonal Antibody [clone: 4C11]
Catalog # ABNOH00002592-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Galactose-1-Phosphate Uridylyltransferase
- Antigen symbol:GALT
- Clonality:Monoclonal
- Clone:4C11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:2592
- Antigen synonyms:UDP glucose hexose 1 phosphate uridylyltransferase|GALT_HUMAN.|Galactose 1 phosphate uridyl transferase|Gal 1 P uridylyltransferase|Galactose 1 phosphate uridylyltransferase
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: GALT
Clonality: Monoclonal
Clone: 4C11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human