Anti-GALT Rabbit Polyclonal Antibody
ABNOH00002592-D01
: Abnova
- Antibody type:Primary
- Antigen name:Galactose-1-Phosphate Uridylyltransferase
- Antigen symbol:GALT
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:2592
- Antigen synonyms:UDP glucose hexose 1 phosphate uridylyltransferase|GALT_HUMAN.|Galactose 1 phosphate uridyl transferase|Gal 1 P uridylyltransferase|Galactose 1 phosphate uridylyltransferase
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: GALT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human