Anti-FANCF Mouse Monoclonal Antibody [clone: 3C4]
Catalog # ABNOH00002188-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Fanconi Anemia Complementation Group F
- Antigen symbol:FANCF
- Clonality:Monoclonal
- Clone:3C4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:2188
- Antigen synonyms:FAF
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: FANCF
Clonality: Monoclonal
Clone: 3C4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human