Ordreindtastning
Denmark
ContactUsLinkComponent
Anti-PTK2B Mouse Monoclonal Antibody [clone: 1F9]
Anti-PTK2B Mouse Monoclonal Antibody [clone: 1F9]
Catalog # ABNOH00002185-M01
Supplier:  Abnova
undefined
Anti-PTK2B Mouse Monoclonal Antibody [clone: 1F9]
Catalog # ABNOH00002185-M01
Supplier:  Abnova

Specifications

  • Antistof type:
    Primary
  • Antigen name:
    Protein Tyrosine Kinase 2 beta
  • Antigen symbol:
    PTK2B
  • Clonality:
    Monoclonal
  • Clone:
    1F9
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Vært:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Krydsadsorption:
    No
  • Gen ID:
    2185
  • Antigen synonymer:
    PTK|PYK2|PKB|FAK2|FADK2|CADTK|RAFTK|CAKB
  • Opbevaringstemperatur:
    Cool
  • Shipping temperature:
    Ice
  • Størrelse:
    100 µg
  • Pakn:
    100 µg

Specifications

About this item

Type: Primary
Antigen: PTK2B
Clonality: Monoclonal
Clone: 1F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human